missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium on activated carbon, 5% Ru, unreduced, ca. 50% moisture, Thermo Scientific Chemicals

£123.00 - £491.00

Chemical Identifiers

CAS 7440-18-8, 7440-44-0, 7732-18-5
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10521261
View Documents
Thermo Scientific Acros
195460250
25 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10531261
View Documents
Thermo Scientific Acros
195461000
100 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-18-8, 7440-44-0, 7732-18-5
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD00011207 MFCD03458417
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
CHEBI:30682
[Ru]

Specifications

Black
1.02
MFCD00011207 MFCD03458417
15, 8433
KJTLSVCANCCWHF-UHFFFAOYSA-N
ruthenium
23950
101.07
Powder
7440-18-8, 7440-44-0, 7732-18-5
Ru
01,983; 05,574
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
[Ru]
101.07
CHEBI:30682
5% (Ru on dry basis)(typically)
Ruthenium on activated carbon, 5% Ruthenium
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ruthenium on carbon, ca. 50% moisture

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.