missing translation for 'onlineSavingsMsg'
Learn More
Ruthenium powder, -325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11381509

Ruthenium powder, -325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11381509
50 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11381509

Brand: Thermo Scientific Alfa Aesar 013994.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]

Specifications

Color Gray
Appearance gray powder
Assay Percent Range 99.9% (Metals basis)
Quantity 50 g
Mesh Size -325 Mesh
Form Powder
Chemical Name or Material Ruthenium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX10K011 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number013994.09,013994.04,013994.18
Lot NumberP25K012 Certificate TypeCertificate of Analysis Date08/04/2024 Catalog Number013994.09,013994.04,013994.18
Lot NumberS13J040 Certificate TypeCertificate of Analysis Date21/06/2023 Catalog Number013994,013994.09,013994.04,013994.18
Lot NumberZ01I018 Certificate TypeCertificate of Analysis Date23/12/2022 Catalog Number013994.09,013994.04,013994.18,013997.09
Lot NumberT25I038 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog Number013994.09,013994.04,013994.18,013997.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ruthenium powder, -325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.