Learn More
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, 98%, Thermo Scientific Chemicals
Chiral catalyst ligand
£16.20 - £615.00
Chemical Identifiers
CAS | 100165-88-6 |
---|---|
Molecular Formula | C48H40P2 |
Molecular Weight (g/mol) | 678.796 |
MDL Number | MFCD00269856 |
InChI Key | IOPQYDKQISFMJI-UHFFFAOYSA-N |
Synonym | r-tol-binap, s-tol-binap, s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, 2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-t-binap, rac-tol-binap, r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, tol-binap, s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-p-tol-binap |
PubChem CID | 4189906 |
IUPAC Name | [1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane |
SMILES | CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329787
|
Thermo Scientific Alfa Aesar
042120.MC |
100 mg | |||||||
11339787
|
Thermo Scientific Alfa Aesar
042120.02 |
0.5 g |
£60.40
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306059
|
Thermo Scientific Alfa Aesar
042120.04 |
2 g |
£161.00
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11316059
|
Thermo Scientific Alfa Aesar
042120.09 |
10 g | |||||||
Description
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl is chiral ligand widely used in asymmetric synthesis. It also used as excellent catalysts for asymmetric hydrogenation of alkenes and some cyclic anhydrides. BINAP is used in organic synthesis for enantioselective transformations catalyzed by its complexes of ruthenium, rhodium, and palladium
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100165-88-6 | |
678.796 | |
IOPQYDKQISFMJI-UHFFFAOYSA-N | |
4189906 | |
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C |
C48H40P2 | |
MFCD00269856 | |
r-tol-binap, s-tol-binap, s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, 2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-t-binap, rac-tol-binap, r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, tol-binap, s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-p-tol-binap | |
[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane |
Specifications
100165-88-6 | |
C48H40P2 | |
r-tol-binap, s-tol-binap, s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, 2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-t-binap, rac-tol-binap, r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, tol-binap, s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-p-tol-binap | |
IOPQYDKQISFMJI-UHFFFAOYSA-N | |
[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane | |
4189906 | |
98% | |
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl |
255°C to 257°C | |
MFCD00269856 | |
Insoluble in water. | |
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C | |
678.796 | |
678.79 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only