missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Methoxypropanol, 97%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 116422-39-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD01632593
InChI Key YTTFFPATQICAQN-BYPYZUCNSA-N
Synonym 2s-2-methoxypropan-1-ol, s-+-2-methoxypropanol, s-2-methoxypropan-1-ol, unii-6725gr28fg, 2-methoxy-1-propanol, 2s, 1-propanol, 2-methoxy-, 2s, 2s-2-methoxy-1-propanol, s-2-methoxy-1-propanol, 2-s-methoxypropan-1-ol, 1-propanol, 2-methoxy-, s
PubChem CID 642402
IUPAC Name (2S)-2-methoxypropan-1-ol
SMILES CC(CO)OC
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10726622
View Documents
Thermo Scientific Acros
331850010
1 mL Glass bottle N/A
N/A
10244751
View Documents
Thermo Scientific Acros
331850025
2.5 mL Glass bottle N/A
N/A
10328653
View Documents
Thermo Scientific
331850100
10 mL Glass bottle N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

116422-39-0
90.12
YTTFFPATQICAQN-BYPYZUCNSA-N
642402
CC(CO)OC
C4H10O2
MFCD01632593
2s-2-methoxypropan-1-ol, s-+-2-methoxypropanol, s-2-methoxypropan-1-ol, unii-6725gr28fg, 2-methoxy-1-propanol, 2s, 1-propanol, 2-methoxy-, 2s, 2s-2-methoxy-1-propanol, s-2-methoxy-1-propanol, 2-s-methoxypropan-1-ol, 1-propanol, 2-methoxy-, s
(2S)-2-methoxypropan-1-ol

Specifications

116422-39-0
100.0
0.9380g/mL
Authentic
C4H10O2
1.4060 to 1.4080
0.938
+ 39.20
YTTFFPATQICAQN-BYPYZUCNSA-N
(2S)-2-methoxypropan-1-ol
642402
97%
(S)-(+)-2-Methoxypropanol, 99%
98.5
Colorless
33°C
96% min. (GC)
CH3CH(OCH3)CH2OH
MFCD01632593
+ 39.20 (20.00°C c=1.66,CHCl3)
2s-2-methoxypropan-1-ol, s-+-2-methoxypropanol, s-2-methoxypropan-1-ol, unii-6725gr28fg, 2-methoxy-1-propanol, 2s, 1-propanol, 2-methoxy-, 2s, 2s-2-methoxy-1-propanol, s-2-methoxy-1-propanol, 2-s-methoxypropan-1-ol, 1-propanol, 2-methoxy-, s
CC(CO)OC
90.12
90.12
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only