Learn More
(S)-4-(4-Aminobenzyl)-2-oxazolidinone, 97%, Thermo Scientific™
£17.60 - £225.00
Chemical Identifiers
CAS | 152305-23-2 |
---|---|
Molecular Formula | C10H12N2O2 |
Molecular Weight (g/mol) | 192.218 |
MDL Number | MFCD03411476 |
InChI Key | WNAVSKJKDPLWBD-VIFPVBQESA-N |
Synonym | s-4-4-aminobenzyl oxazolidin-2-one, s-4-4-aminobenzyl-2-oxazolidinone, s-4-4-aminobenzyl-2 1h-oxazolidinone, s-4-4-aminobenzyl-1,3-oxazolidin-2-one, s-4-4-amino-benzyl-oxazolidine-2-one, 4s-4-4-aminophenyl methyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-4-aminophenyl methyl-, 4s, s-4-4-aminobenzyl-1,3-oxozolidin-2-one, 4s-4-4-aminophenyl methyl-2-oxazolidinone, s-4-4'-aminobenzyl-1,3-oxazolidin-2-one |
PubChem CID | 7099156 |
IUPAC Name | (4S)-4-[(4-aminophenyl)methyl]-1,3-oxazolidin-2-one |
SMILES | C1C(NC(=O)O1)CC2=CC=C(C=C2)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15472438
|
Thermo Scientific Alfa Aesar
H56017.03 |
1 g |
£17.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15492438
|
Thermo Scientific Alfa Aesar
H56017.14 |
25 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications(S)-4-(4-Aminobenzyl)-2-(1H)-oxazolidinone is an intermediate of zolmitriptan (sc-220415) which acts as a novel inhibitor of the cytochrome P-450 enzyme aromatase.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store at -20°C. Incompatible with oxidizing agents. Keep container tightly closed.
Chemical Identifiers
152305-23-2 | |
192.218 | |
WNAVSKJKDPLWBD-VIFPVBQESA-N | |
7099156 | |
C1C(NC(=O)O1)CC2=CC=C(C=C2)N |
C10H12N2O2 | |
MFCD03411476 | |
s-4-4-aminobenzyl oxazolidin-2-one, s-4-4-aminobenzyl-2-oxazolidinone, s-4-4-aminobenzyl-2 1h-oxazolidinone, s-4-4-aminobenzyl-1,3-oxazolidin-2-one, s-4-4-amino-benzyl-oxazolidine-2-one, 4s-4-4-aminophenyl methyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-4-aminophenyl methyl-, 4s, s-4-4-aminobenzyl-1,3-oxozolidin-2-one, 4s-4-4-aminophenyl methyl-2-oxazolidinone, s-4-4'-aminobenzyl-1,3-oxazolidin-2-one | |
(4S)-4-[(4-aminophenyl)methyl]-1,3-oxazolidin-2-one |
Specifications
152305-23-2 | |
C10H12N2O2 | |
s-4-4-aminobenzyl oxazolidin-2-one, s-4-4-aminobenzyl-2-oxazolidinone, s-4-4-aminobenzyl-2 1h-oxazolidinone, s-4-4-aminobenzyl-1,3-oxazolidin-2-one, s-4-4-amino-benzyl-oxazolidine-2-one, 4s-4-4-aminophenyl methyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-4-aminophenyl methyl-, 4s, s-4-4-aminobenzyl-1,3-oxozolidin-2-one, 4s-4-4-aminophenyl methyl-2-oxazolidinone, s-4-4'-aminobenzyl-1,3-oxazolidin-2-one | |
WNAVSKJKDPLWBD-VIFPVBQESA-N | |
(4S)-4-[(4-aminophenyl)methyl]-1,3-oxazolidin-2-one | |
7099156 | |
97% |
107°C to 111°C | |
MFCD03411476 | |
Soluble in dimethyl sulfoxide and methanol. | |
C1C(NC(=O)O1)CC2=CC=C(C=C2)N | |
192.218 | |
192.22 | |
(S)-4-(4-Aminobenzyl)-2-oxazolidinone |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures