Learn More
Heidolph Instruments S-Box Package 1
The S-Box Package 1 includes additional accessories to complete an existing S-Biosystem incubation system.
£3595.00
Specifications
Includes 1 S-box, 1 Tool Kit, 4 inner boxes, 4 water bags, 4 front covers
View More Specs
Products 4
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
17848000
View Documents
Heidolph Instruments
400-05000-00
Green N/A N/A N/A
17858000
View Documents
Heidolph Instruments
400-05001-00
Orange N/A N/A N/A
17868000
View Documents
Heidolph Instruments
400-05002-00
Blue N/A N/A N/A
17878000
View Documents
Heidolph Instruments
400-05003-00
Violet N/A N/A N/A
Description
Description
  • Each S-box is a stand-alone miniature incubator.
  • Inside the S-box there is room for one standard multi-well plate, or one cell culture bottle (up to T75).
  • The complete interior (inner box) can be purchased as a sterilized exchangeable element in a sterile outer packaging.
  • Your culture is prevented from drying out by a sachet of sterilized water (Waterbag), which is enough for weeks.
  • To avoid mix-up, the front cover of the S-box is available in different colors.
Specifications
Specifications
1 S-box, 1 Tool Kit, 4 inner boxes, 4 water bags, 4 front covers
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Heidolph Instruments S-Box Package 1