Learn More
Heidolph Instruments S-Box Package 2
The S-Box Package 2 includes additional accessories to complete an existing S-Biosystem incubation system.
£4015.00
Specifications
Includes 1 S-box, 1 tool kit, 8 inner boxes, 8 water bags, 4 front covers
View More Specs
Products 4
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
17808000
View Documents
Heidolph Instruments
400-03000-00
Green N/A N/A N/A
17818000
View Documents
Heidolph Instruments
400-03001-00
Orange N/A N/A N/A
17828000
View Documents
Heidolph Instruments
400-03002-00
Blue N/A N/A N/A
17838000
View Documents
Heidolph Instruments
400-03003-00
Violet N/A N/A N/A
Description
Description
  • Each S-box is a stand-alone miniature incubator.
  • Inside the S-box there is room for one standard multi-well plate, or one cell culture bottle (up to T75).
  • The complete interior (inner box) can be purchased as a sterilized exchangeable element in a sterile outer packaging.
  • Your culture is prevented from drying out by a sachet of sterilized water (Waterbag), which is enough for weeks.
  • To avoid mix-up, the front cover of the S-box is available in different colors.
Specifications
Specifications
1 S-box, 1 tool kit, 8 inner boxes, 8 water bags, 4 front covers
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Heidolph Instruments S-Box Package 2