missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-N-BOC-α-Ethylalanine, 98%, 98% ee, Thermo Scientific Chemicals
£92.20 - £268.00
Chemical Identifiers
CAS | 151171-11-8 |
---|---|
Molecular Formula | C10H19NO4 |
Molecular Weight (g/mol) | 217.27 |
MDL Number | MFCD12031697 |
InChI Key | SHZXLTCEPXVCSV-JTQLQIEISA-N |
Synonym | boc-l-isovaline, s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, s-2-tert-butoxycarbonylamino-2-methylbutanoic acid, n-1,1-dimethylethyl oxy carbonyl-d-isovaline, s-2-methyl-2-tert-butoxycarbonylamino butyric acid, 2s-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid |
PubChem CID | 14284792 |
IUPAC Name | (2S)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
SMILES | CCC(C)(NC(=O)OC(C)(C)C)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11955521
|
Thermo Scientific Acros
441131000 |
100 mg |
£92.20
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11965521
|
Thermo Scientific Acros
441135000 |
500 mg |
£268.00
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
151171-11-8 | |
217.27 | |
SHZXLTCEPXVCSV-JTQLQIEISA-N | |
14284792 | |
CCC(C)(NC(=O)OC(C)(C)C)C(O)=O |
C10H19NO4 | |
MFCD12031697 | |
boc-l-isovaline, s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, s-2-tert-butoxycarbonylamino-2-methylbutanoic acid, n-1,1-dimethylethyl oxy carbonyl-d-isovaline, s-2-methyl-2-tert-butoxycarbonylamino butyric acid, 2s-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid | |
(2S)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
Specifications
104°C to 112°C | |
97.5 | |
White to Yellow | |
C10H19NO4 | |
MFCD12031697 | |
boc-l-isovaline, s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, s-2-tert-butoxycarbonylamino-2-methylbutanoic acid, n-1,1-dimethylethyl oxy carbonyl-d-isovaline, s-2-methyl-2-tert-butoxycarbonylamino butyric acid, 2s-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid | |
CCC(C)(NC(=O)OC(C)(C)C)C(O)=O | |
12 | |
14284792 | |
98%,98% ee | |
(S)-N-BOC-α-Ethylalanine, 98% ee |
151171-11-8 | |
100.0 | |
98% | |
Authentic | |
Glass Bottle | |
SHZXLTCEPXVCSV-JTQLQIEISA-N | |
(2S)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid | |
217.27 | |
217.27 | |
Powder |
Safety and Handling
Product Identifier
- (S)-N-BOC-alpha-Ethylalanine
RUO – Research Use Only