missing translation for 'onlineSavingsMsg'
Learn More

s-Trioxane, 99+%, Thermo Scientific Chemicals

£13.00 - £71.36

Chemical Identifiers

CAS 110-88-3
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00006563
InChI Key BGJSXRVXTHVRSN-UHFFFAOYSA-N
Synonym s-trioxane, trioxymethylene, metaformaldehyde, triformol, formaldehyde, trimer, aldeform, formagene, marvosan, trioxan, sym-trioxane
PubChem CID 8081
ChEBI CHEBI:38043
IUPAC Name 1,3,5-trioxane
SMILES C1OCOCO1
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10407160
View Documents
Thermo Scientific Acros
140290050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10227313
View Documents Promotion Details
Thermo Scientific Acros
140291000
100 g Plastic bottle
£14.07
100g
Please to purchase this item. Need a web account? Register with us today!
10636152
View Documents Promotion Details
Thermo Scientific Acros
140295000
500 g Plastic bottle
£26.73
500g
Please to purchase this item. Need a web account? Register with us today!
10021610
View Documents Promotion Details
Thermo Scientific Acros
140290020
2 kg Plastic bottle
£71.36
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

110-88-3
90.08
BGJSXRVXTHVRSN-UHFFFAOYSA-N
8081
1,3,5-trioxane
C3H6O3
MFCD00006563
s-trioxane, trioxymethylene, metaformaldehyde, triformol, formaldehyde, trimer, aldeform, formagene, marvosan, trioxan, sym-trioxane
CHEBI:38043
C1OCOCO1

Specifications

110-88-3
Colorless to White
114.0°C to 116.0°C
Authentic
C3H6O3
19,381
1.17
Solubility in water: soluble in water: 221g/L (25°C). Other solubilities: soluble in alcohols,ether,ketones,acetone,,chlorinatid and aromatic hydrocarbons,and,other organic solvents,slightly soluble in pentane and petr.ether
BGJSXRVXTHVRSN-UHFFFAOYSA-N
1,3,5-trioxane
8081
90.08
Crystalline Flakes or Crystals
61.0°C to 62.0°C
1.1700g/mL
45°C
99% min. (GC)
MFCD00006563
15,9906
s-trioxane, trioxymethylene, metaformaldehyde, triformol, formaldehyde, trimer, aldeform, formagene, marvosan, trioxan, sym-trioxane
0115.0°C
C1OCOCO1
90.08
CHEBI:38043
99+%
s-Trioxane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Suspected of damaging the unborn child.
Flammable solid.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-812-5

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only