missing translation for 'onlineSavingsMsg'
Learn More
Saccharin, 98+%, Thermo Scientific Chemicals Product Code.: 10235340

Saccharin, 98+%, Thermo Scientific Chemicals

Product Code. 10235340
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10235340

Brand: Thermo Scientific Acros 149005000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-07-2
Molecular Formula C7H5NO3S
Molecular Weight (g/mol) 183.18
InChI Key CVHZOJJKTDOEJC-UHFFFAOYSA-N
Synonym saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinoseShow MoreShow Less
PubChem CID 5143
ChEBI CHEBI:32111
IUPAC Name 1,1-dioxo-1,2-benzothiazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)NS2(=O)=O

Specifications

Melting Point 226.0°C to 230.0°C
Color White
Loss on Drying 1% (105°C, 2 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Quantity 500 g
Beilstein 27,168
Merck Index 15,8445
Solubility Information Solubility in water: 3.3g/L (20°C). Other solubilities: 1g/31mL alcohol - 1g/12mL acetone, 1g/50mL glycerol, soluble in amyl acetate and ethyl acetate, slightly soluble in chloroform and ether, soluble in dil. solution of ammonia
Formula Weight 183.18
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Saccharin
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460629 Certificate TypeCertificate of Analysis Date27/02/2024 Catalog Number149000025,149001000,149005000
Lot NumberA0440985 Certificate TypeCertificate of Analysis Date09/10/2023 Catalog Number149000025,149001000,149005000
Lot NumberA0403075 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number149000025,149001000,149005000
Lot NumberA0411180 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number149000025,149001000,149005000
Lot NumberA0409553 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number149000025,149001000,149005000
Product Identifier
  • Saccharin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Saccharin, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.