missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Safe Lock Micro Test Tube
GREENER_CHOICE

Use these polypropylene micro tubes for a variety of applications. Eppendorf™ Safe Lock Micro Test Tubes feature hinged Eppendorf Safe-Lock lids for outstanding protection against unintentional opening during incubation and storage.

Brand:  Eppendorf™ 0030120221

82.09 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.


 View more versions of this product

Product Code. 10663981

  • £90.06 / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage
  • Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without mold release agents, softeners or biocides that could have a negative effect on bioassays
  • g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Precise lid sealing for minimal evaporation rates during long-term storage
  • Large frosted lid and surface on the side for easy labeling
  • Guaranteed functionality from -86 to +100°C
  • Autoclavable when open (121°C, 20 min.)
  • Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur quality for guaranteed purity
  • Available in Eppendorf Protein and DNA LoBind material for maximum recovery of valuable samples
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 mL
Polypropylene
–86 to 100°C
Blue
1000/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Safe Lock Micro Test Tube > Color: Blue; Capacity: 2.0mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.