Learn More
Brady™ SafeKey Compact Padlocks - Nylon with 25 mm Plastic Shackle, Keyed Different
A small and lightweight range of padlocks suitable for energy isolation programs or in small spaces.
£15.70 - £93.85
Specifications
Product Type | Keyed Padlocks |
---|---|
Key Type | Keyed Different |
Material | Nylon |
Diameter (English) Shackle | 0.19 in. |
Diameter (Metric) Shackle | 4.7 mm |
Product Code | Brand | Quantity | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Color | Price | Quantity & Availability | ||||
17190241
|
Brady™
150180 |
1 Ea. | Red |
£15.70
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17100251
|
Brady™
150181 |
1 Ea. | Yellow |
£18.50
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17110251
|
Brady™
150182 |
1 Ea. | Green |
£16.05
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17120251
|
Brady™
150183 |
1 Ea. | Blue |
£15.70
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17130251
|
Brady™
150184 |
1 Ea. | Black |
£15.70
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17140251
|
Brady™
150185 |
1 Ea. | Orange |
£15.70
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17150251
|
Brady™
150186 |
1 Ea. | Purple |
£15.70
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17160251
|
Brady™
150187 |
1 Ea. | Brown |
£15.70
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17170251
|
Brady™
150188 |
1 Ea. | White |
£15.70
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17170261
|
Brady™
150207 |
6 Pk. | Red |
£92.25
Pack of 6 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17180261
|
Brady™
150208 |
6 Pk. | Yellow |
£93.85
Pack of 6 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17190261
|
Brady™
150209 |
6 Pk. | Green |
£92.25
Pack of 6 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17100271
|
Brady™
150210 |
6 Pk. | Blue |
£92.25
Pack of 6 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17110271
|
Brady™
150211 |
6 Pk. | Black |
£92.25
Pack of 6 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17120271
|
Brady™
150212 |
6 Pk. | Orange |
£92.25
Pack of 6 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
17130271
|
Brady™
150213 |
6 Pk. | Purple |
£92.25
Pack of 6 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
17140271
|
Brady™
150214 |
6 Pk. | Brown |
£92.25
Pack of 6 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
17150271
|
Brady™
150215 |
6 Pk. | White |
£92.25
Pack of 6 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
Description
- Compliments the Brady™ SafeKey Padlock range and have the same features
- The new designed and innovative keying mechanism provides you much more unique key possibilities than a conventional standard safety padlock, without sacrificing a secure lockout
- The unique lock mechanism is insulated and non-conductive to protect workers from shocks when key is inserted
- The lock mechanism has 6 precision steel tumbler blades for a high number of key differences and tamper resistance
- Locks are compact and lightweight and resist impact, temperature, chemicals and corrosion
- Their non-conductive, non-sparking body features a ribbed design for easier gripping and handling
- The colour-matched key and lock body allows a fast identification in the application
- 1 key is supplied with each lock
- Custom keying, engraving, charting, Master Key and Grand Master Key are available
Specifications
Keyed Padlocks | |
Nylon | |
4.7 mm | |
25 mm | |
6 | |
1 in. | |
26 mm |
Keyed Different | |
0.19 in. | |
1 in. | |
Plastic | |
1.3 in. | |
33 mm |