missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Safety Bottle Carrier
The Safety Bottle Carrier provides a safe and reliable way to transport a variety of bottled reagents in your lab.
Brand: Fisherbrand™ 120913F
This item is not returnable.
View return policy
Description
- Robust Design
- Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
- Handle can be pressed down to either side for easier loading and unloading
- Centering ring stabilizes bottle in the center of carrier for safer, steadier transport
- Carriers come in four colors with designated labeling areas for easy identification
- Made of chemical resistant thermoplastic elastomer - PP blend
Specifications
1 Safety Bottle Carrier vessel and attachable handle | |
Safety Bottle Carrier, 4.5 L, Blue | |
Not Autoclavable | |
Bottles | |
1 | |
None | |
Non-sterile | |
4.5 L bottles | |
-20 to 60°C | |
18.3 cm | |
18.3 cm | |
.25 kg |
Bottle Holder | |
1 x 1 | |
Blue | |
Polypropylene | |
Cylindrical | |
No | |
Transportation of bottled regeants | |
4.5 L | |
18.3 cm | |
26.7 cm (40.6 cm Height, Handle Extended) | |
18.3 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction