missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT HPLC Safety Caps, V2.0, with Shut-Off
SafetyCap is designed to safety extract solvents when working with any HPLC/UHPLC system. The SCAT Safety Caps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours.
£143.00 - £253.00
Specifications
Type | Safety Cap |
---|---|
For Use With (Application) | HPLC Systems |
For Use With (Equipment) | Lab Bottles |
Material | PFA, PTFE, PPS |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
16423809
|
SCAT
307919 |
2 fittings, 1 air valve, 2 shut-offs |
£143.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
17172576
|
SCAT
307419 |
4 x shut-off, 4 x PFA-fittings 3.2 mm OD, and air valve |
£194.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
16423789
|
SCAT
307219 |
2 fittings, 1 air valve, shut-off (combined) |
£143.00
Each |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||||
16433809
|
SCAT
307920 |
3 fittings, 1 air valve, 3 shut-offs |
£154.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
16443799
|
SCAT
307319 |
3 fittings, 1 air valve, 2 shut-offs (combined) |
£154.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
16483799
|
SCAT
307519 |
6 fittings, 1 air valve, 6 shut-offs |
£253.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
Description
- Luer Lock Adapter
- Improved fitting design for easier connection
- Constructed of medical grade PTFE, PFA fittings, and a PPS screw cap
- Includes space for individually labeling
- Autocalvable up to 200°C
- Stable construction for improved handling
Specifications
Safety Cap | |
HPLC Systems | |
Lab Bottles | |
PFA, PTFE, PPS |