Learn More
Fisherbrand™ Safety Cover for Dry Block Heaters
For use with Fisherbrand™ Dry Block Heaters.
Brand: Fisherbrand™
Additional Details : Weight : 0.30000kg
Description
- Made from tough clear acrylic for maximum visibility
- Prevents accidental touching of a hot block or contamination of samples from splashes
Spécification
Fisherbrand™ 4 Block System | |
Preventing Burns or Contamination | |
85 mm |
Safety Cover | |
Acrylic |