missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Safety Cover for Dry Block Heaters
For use with Fisherbrand™ Dry Block Heaters.
£101.07 - £115.54
Specifications
Type | Safety Cover |
---|---|
For Use With (Application) | Preventing Burns or Contamination |
Material | Acrylic |
Height (Metric) | 85 mm |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17561896
|
Fisherbrand™
|
Fisherbrand™ 1 Block System |
£101.07
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17571896
|
Fisherbrand™
|
Fisherbrand™ 2 Block System |
£109.18
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17581896
|
Fisherbrand™
|
Fisherbrand™ 4 Block System |
£115.54
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
Description
- Made from tough clear acrylic for maximum visibility
- Prevents accidental touching of a hot block or contamination of samples from splashes
Specifications
Safety Cover | |
Preventing Burns or Contamination | |
Acrylic | |
85 mm |