Learn More
BTX™ Safety Dome for Electroporation Cuvettes
Ensure safe delivery of HV electrical pulses to cuvettes and chambers with the Safety Dome for Electroporation Cuvettes.
Brand:  BTX™ 452021
 View more versions of this product
Product Code. 15497360
  • £1175.00 / Each
Explore more special offers
Description
Description
  • Safety dome plugs directly into generator
  • Holds two cuvettes
  • Safety swing shield secures cuvettes in holder
Specifications
Specifications
Electroporation
BTX Electroporation
3000 VDC, 250 VAC
CE Marking, ETL Marking (UL, CSA), EU RoHS, WEEE
41°F to 104°F
5°C to 40°C
2.3 lb.
Non-sterile
Retractable Sheath Banana Plug
BTX Gemini X2, ECM630, ECM830
Electroporation Safety Stands
Safety Dome for the Gemini X2
Safety Dome Only
10 μsec. to 10 sec.
1 Ea.
1 kg
-10°C to +70°C
Exponential Decay Wave, Square Wave
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BTX™ Safety Dome for Electroporation Cuvettes > Safety Dome; For use with: Gemini X2