missing translation for 'onlineSavingsMsg'
Learn More

Notrax Safety Stance™ High Quality Anti-Fatigue Matting

Notrax Safety Stance™ High Quality Anti-Fatigue Matting - High quality anti-fatigue matting for use in areas where there can be frequent oil spilling. Drainage holes and raised studs provide aeration and allow fluids and debris to fall through, leaving a dry and tidy top surface.

Brand:  Notrax 549S2640OB

 View more versions of this product

Product Code. 17732198

  • £175.00 / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High quality anti-fatigue matting for use in areas where there can be frequent oil spilling
  • Drainage holes and raised studs provide aeration and allow fluids and debris to fall through, leaving a dry and tidy top surface
  • Made of 100% nitrile rubber compound, designed to withstand industrial oils and harsh chemicals
  • Suited for multi-shift operations
  • Ergonomic benefit derived from 22 mm thick resilient worker platform
  • Highly visible nitrile rubber orange colored borders included on 3 sides for added safety
  • Aggressive slip resistant surface
  • Cleaning with detergent and waterjet is recommended for extended service life
  • Standard size mats are loose lay and require no installation
  • Non-shifting
  • Free of silicone therefore safe for vehicle painting facilities
Specifications

Specifications

Black/Orange
102 cm
Industrial and Commercial Matting
3.3 ft.
Natural Rubber
66 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.