missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Safety Stance™ High Quality Anti-Fatigue Matting
Notrax Safety Stance™ High Quality Anti-Fatigue Matting - High quality anti-fatigue matting for use in areas where there can be frequent oil spilling. Drainage holes and raised studs provide aeration and allow fluids and debris to fall through, leaving a dry and tidy top surface.
Brand: Notrax 549S3124OB
This item is not returnable.
View return policy
Description
- High quality anti-fatigue matting for use in areas where there can be frequent oil spilling
- Drainage holes and raised studs provide aeration and allow fluids and debris to fall through, leaving a dry and tidy top surface
- Made of 100% nitrile rubber compound, designed to withstand industrial oils and harsh chemicals
- Suited for multi-shift operations
- Ergonomic benefit derived from 22 mm thick resilient worker platform
- Highly visible nitrile rubber orange colored borders included on 3 sides for added safety
- Aggressive slip resistant surface
- Cleaning with detergent and waterjet is recommended for extended service life
- Standard size mats are loose lay and require no installation
- Non-shifting
- Free of silicone therefore safe for vehicle painting facilities
Specifications
Black/Orange | |
315 cm | |
Industrial and Commercial Matting |
10.3 ft. | |
Natural Rubber | |
97 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction