missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Cabinet for Acids and Alkalis SL-CLASSIC
Safe and approved storage of aggressive, non flammable hazardous materials in working areas.
Brand: asecos™ 3060600130607
2946.32 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Alert:
For indoor use only
Description
Safe and approved storage of aggressive, non flammable hazardous materials in working areas
Function / construction
- Robust construction and longevity: cabinet body made of powder-coated sheet steel, safety elements assembled outside the storage compartment for increased protection against corrosion
- Two in one: two hermetically separated storage compartments allow the isolated storage of acids and bases in one cabinet
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- ventilation: integrated metal-free air ducts ready for connection to a technical exhaust system with only one extraction air inlet
Certificates
- type tested in accordance with EN 14727
- EN 16121/16122 conformity
- GS certified
Services
Five years
Specifications
DIN EN 16121/16122 conformity, GS certified | |
603 mm | |
570 x 485 x 1869 mm | |
Wing | |
1965 mm | |
SL.196.120.MV | |
8 | |
Melamine resin laminated | |
1197 mm | |
Light Gray | |
Cabinet for acids and alkalis |
8 x pull-out shelf (panel melamine resin-coated/PP) | |
570 mm | |
603 x 1197 x 1965 mm | |
Safe and approved storage of aggressive, non fla mmable hazardous materials in working areas | |
1865 mm | |
2 | |
Pull-out | |
271.7 kg | |
485 mm | |
Chipboard, Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction