Learn More
SCAT™ Safety Waste Cap 'LISA', V3.0 Electronic Level Control Extension
With the extension Level Control, you can use your LISA to monitor the fill level of your disposal container at any time.
Brand:  SCAT™ 350122
Additional Details : Weight : 0.55800kg
 View more versions of this product
Product Code. 17873530
  • £108.00 / Pack of 3
Estimated Shipment: 13-08-2024
to see stock.
Add to basket
Description
Description
LISA is the first modular system for HPLC waste, providing occupational safety and environmental protection with proven SCAT™ technology and maximum flexibility. Simply add connections and components as required so you can collect solvents, acids and alkalis safely at any time.
  • Simply unscrew the blind plug of the LISA, insert the level control, and connect cable to the signal box
  • Permanently displays the fill level in your disposal container and alerts when reached as soon as it is full
  • Available in electrically conductive and non-electrically conductive versions
Specifications
Specifications
HPLC Systems
Level Control
PTFE
SCAT™ Safety Waste Cap 'LISA', V3.0