Learn More
SCAT™ HPLC Safety Caps, V2.0
GREENER_CHOICE
These HPLC Safety Caps allow you to extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapors. Complete with integrated air valve for pressure equalization.
Brand:  SCAT™ 307909-PP
 View more versions of this product
Product Code. 18686488
  • £106.00 / 1 sheet
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
SCAT™ Safety Caps V2.0 provide maximum safety for your HPLC solvent supply.
  • High quality materials ensure maximum safety for lab and staff
  • 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
  • Stable construction for highest safety and easy handling
  • Chemically resistant against aggressive organic solvents
  • Attractive design and clear arrangement of connections
  • Improved design for easy connection of capillaries in various colors
  • Free air valve with change indicator included with every Safety Cap
Specifications
Specifications
HPLC Systems
Safety Cap II: 1 x air valve, 1 x change indicator, 2 x PP fitting 1.6/2.3/3.3 mm OD, 1 x PP blind plug, 1 x labelling field
GL45
Lab Bottle
Polypropylene
Safety Cap