missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint Gobain Life Sciences Chemware™ Bendable PTFE Stirring Rod
Chemware™ Bendable PTFE Stirring Rod contains a fully enclosed steel insert for stiffness, yet can be bent to various shapes for use as powered stirrers.
Marque: Saint Gobain Life Sciences D1069089
Code : 03
Informations supplémentaires : Weight : 0.01000kg
Description
- Heat resistant to 288°C (550°F)
- Completely inert to highly corrosive reagents
- Anti-stick properties facilitate cleaning
- Unbreakable
Spécification
Fully enclosed steel insert | |
12 in. | |
Stirring Rod | |
PTFE |
6 mm | |
0.25 in. | |
1 Ea. | |
30 mm |