missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint Gobain Life Sciences Chemware™ Bendable PTFE Stirring Rod
Chemware™ Bendable PTFE Stirring Rod contains a fully enclosed steel insert for stiffness, yet can be bent to various shapes for use as powered stirrers.
£110.73 - £174.00
Specifications
Material | PTFE |
---|---|
Quantity | 1 Ea. |
Type | Stirring Rod |
Includes | Fully enclosed steel insert |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
12318219
|
Saint Gobain Life Sciences
D1069087 |
6 mm | 20 mm |
£116.00
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
12338219
|
Saint Gobain Life Sciences
D1069089 |
6 mm | 30 mm |
£174.00
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
12348219
|
Saint Gobain Life Sciences
D1069091 |
8 mm | 40 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Heat resistant to 288°C (550°F)
- Completely inert to highly corrosive reagents
- Anti-stick properties facilitate cleaning
- Unbreakable
Specifications
PTFE | |
1 Ea. | |
Stirring Rod | |
Fully enclosed steel insert |