missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Chemware™ Bendable PTFE Stirring Rod

Chemware™ Bendable PTFE Stirring Rod contains a fully enclosed steel insert for stiffness, yet can be bent to various shapes for use as powered stirrers.

Brand:  Saint Gobain Life Sciences D1069087

 View more versions of this product

Product Code. 12318219

  • £116.00 / Each
Estimated Shipment: 15-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Heat resistant to 288°C (550°F)
  • Completely inert to highly corrosive reagents
  • Anti-stick properties facilitate cleaning
  • Unbreakable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.25 in.
8 in.
20 mm
1 Ea.
Fully enclosed steel insert
6 mm
PTFE
Stirring Rod
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers