Learn More
Saint Gobain Life Sciences Chemware™ PTFE Boiling Stones
Use these PTFE chips to promote gentle, efficient boiling and help prevent overheating of highly volatile liquids. Chemware™ PTFE Boiling Stones are not affected by acids, hydrocarbons, ketones, esters, alcohols or alkalis.
Brand:  Saint Gobain Life Sciences D1069103
Product Code. 10296104
  • £77.73 / Each
Estimated Shipment: 10-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use these PTFE chips to promote gentle, efficient boiling and help prevent overheating of highly volatile liquids. Saint Gobain Performance Plastics™ Chemware™ PTFE Boiling Stones are not affected by acids, hydrocarbons, ketones, esters, alcohols or alkalis.
  • Use these PTFE chips to promote gentle, efficient boiling and help prevent overheating of highly volatile liquids.
  • Saint Gobain Life Sciences Chemware™ PTFE Boiling Stones are not affected by acids, hydrocarbons, ketones, esters, alcohols or alkalis.
  • Plastic Bottle
At temperatures above 260°C (500°F), a few chemicals at high concentration have been reported reactive to PTFE. These include 80% NaOH or KOH, metal hydrides (such as boranes), aluminum chloride, ammonia and certain amines and imines. Also, slow oxidative attack by 70% nitric acid under pressure at 250°C (480°F) has been observed. Special testing is required when extremes of reducing or oxidizing conditions are approached.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Boiling Stone
Boiling Stones
16 oz. (450 g)
PTFE
Distillation Columns
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Saint Gobain Life Sciences Chemware™ PTFE Boiling Stones > Chemware™ PTFE Boiling Stones; Quantity: 16 oz. (450 g)