Learn More
Saint Gobain Life Sciences Chemware™ PTFE PFA Griffin Beakers
Chemware™ PTFE PFA Griffin Beakers ideal for ultrapure applications.
£51.40 - £83.27
Specifications
Color Transparent
Material PTFE
Type Beaker
Autoclavable Autoclavable
Disposable Reusable
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10787175
View Documents
Saint Gobain Life Sciences
D1069536
50 mL
£51.40
Each
Estimated Shipment: 09-04-2025
to see stock.
Add to basket
12356988
View Documents
Saint Gobain Life Sciences
D1069537
125 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Designed for rapid heat transfer
  • Can be heated in a microwave or directly on a hot plate up to 500°F (260°C)
  • Transparent
  • Graduated for easy measuring
  • Autoclavable
Specifications
Specifications
Transparent
Beaker
Reusable
Griffin
PTFE
Autoclavable
Yes