Learn More
Salicylic Acid, 99+%, Thermo Scientific Chemicals
£49.35 - £83.62
Chemical Identifiers
CAS | 69-72-7 |
---|---|
Molecular Formula | C7H6O3 |
Molecular Weight (g/mol) | 138.12 |
MDL Number | MFCD00002439 |
InChI Key | YGSDEFSMJLZEOE-UHFFFAOYSA-N |
Synonym | salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone |
PubChem CID | 338 |
ChEBI | CHEBI:16914 |
IUPAC Name | 2-hydroxybenzoic acid |
SMILES | OC(=O)C1=CC=CC=C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10367170
|
Thermo Scientific Acros
147700010 |
1 kg | Plastic bottle |
£49.35
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10543504
|
Thermo Scientific Acros
147700025 |
2.5 kg | Plastic bottle |
£83.62
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69-72-7 | |
138.12 | |
YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
338 | |
2-hydroxybenzoic acid |
C7H6O3 | |
MFCD00002439 | |
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone | |
CHEBI:16914 | |
OC(=O)C1=CC=CC=C1O |
Specifications
69-72-7 | |
211.0°C (20.0 mmHg) | |
99+% | |
MFCD00002439 | |
15, 8469 | |
YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
2-hydroxybenzoic acid | |
338 | |
138.12 | |
157°C | |
Crystalline Powder |
158.0°C to 161.0°C | |
White | |
C7H6O3 | |
10, 43 | |
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone | |
OC(=O)C1=CC=CC=C1O | |
138.12 | |
CHEBI:16914 | |
≥99% | |
Authentic | |
Salicylic acid |
Safety and Handling
GHS H Statement
Harmful if swallowed. Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Do not eat, drink or smoke when using this product.
Danger
- Salicylic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only