missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

£81.41 - £293.46

Chemical Identifiers

CAS 13569-60-3
Molecular Formula Cl3H2O13Sm
Molecular Weight (g/mol) 466.71
MDL Number MFCD00049769
InChI Key ONSMNAINZIBBGR-UHFFFAOYSA-K
Synonym samarium 3+ ;triperchlorate, samarium iii perchlorate solution, trihyperchloric acid samarium iii salt, samarium iii perchlorate w/w aqueous solution, reagent grade
IUPAC Name samarium(3+) hydrate triperchlorate
SMILES O.[Sm+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15436335
SDS View Documents
Thermo Scientific Alfa Aesar
040567.09
10 g
£81.41
10g
Please to purchase this item. Need a web account? Register with us today!
15446335
SDS View Documents
Thermo Scientific Alfa Aesar
040567.18
50 g
£293.46
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H271-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.

missing translation for 'einecsNumber' : 236-986-6

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Samarium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.