Learn More
Novus Biologicals™ Sambucus Nigra Lectin [Cy5]
Brand: Novus Biologicals™ CL-1305-NB
245.60 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
1 mg/mL | |
10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.1 mM Ca^2+, 0.01 mM Mn^2+, protein stabilizer. | |
Store at 4°C in the dark. | |
Glycobiology |
Immunocytochemistry/Immunofluorescence | |
1 mg | |
A precipitate may form during storage. This does not have a significant adverse effect on the product. If a precipitate forms upon long-term storage at 4°C, warm to 37C and centrifuge before use. The recommended concentration range for use is 5-20 μg/mL. | |
Sambucus Nigra Lectin |
Product Content Correction
Product Title
Novus Biologicals™ Sambucus Nigra Lectin [Cy5] > 1 mg
Spot an opportunity for improvement?Share a Content Correction