Learn More
Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes
GREENER_CHOICE
Perform precise dispensing of volumes from 20 to 400 μL by using these exact volume transfer pipettes. Made of shatterproof, nontoxic polyethylene.
Brand:  Thermo Scientific™ 930NL
 View more versions of this product
Product Code. 15367903
  • £27.95 / Pack of 500
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Perform precise dispensing of volumes from 20 to 400 μL by using Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.
  • Shatterproof, nontoxic polyethylene
  • Nonsterile
  • Low affinity surface
  • Freezer ready
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50μL Exact Volume Transfer Pipette
LDPE
No
50 μL
Exact Volume Transfer Pipet
2.5 mm
Non-sterile
Bulk 500/Pk 10Pk/Cs 5000/Cs
5.9 cm
Single-use