missing translation for 'onlineSavingsMsg'
Learn More

KARTELL Sample Container

Sample Container has a screw cap, making it ideal for sample collection, transport and storage.

Brand:  KARTELL 0167300

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10274041

  • £85.35 / Each
Estimated Shipment: 30-10-2024
to see stock.

Description

Description

  • With screw cap (PFA) for sample collection, transport and storage.
  • PFA trace analysis labware maintains its stability at temperatures from -200°C to
    +260°C (-328°F to +500°F).
  • PFA is manufactured without metals such as calcium, aluminum, iron, magnesium, nickel, copper, manganese or zinc. These metals can leach into samples from other containers, and contaminate samples.
  • PFA is inert with most reagents, including nitric and hydrochloric acid routinely used in cleaning trace analysis labware. This stability significantly reduces the risk of cross-contamination.
  • PFA containers often extend the stability of low (ppb) concentration standards. Such stability reduces the time and cost of renewing trace analysis standard solutions.
  • Translucency is important for making accurate measurements using volumetric labware.
  • Conventional labware for trace analysis requires time-consuming and expensive cleaning. The hydrophobic and antiadhesive properties of the exceptionally smooth-surfaced PFA labware simplify the cleaning process, making it ideal for trace analysis.
Specifications

Specifications

Specimen Container
No
Cylindrical
2.03 oz.
Screw Cap
Yes
PFA
Non-sterile
60 mL
Certified Quality
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers