missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL Sample Container
Sample Container has a screw cap, making it ideal for sample collection, transport and storage.
£71.05 - £111.00
Specifications
Product Type | Specimen Container |
---|---|
Closure Included | Yes |
Graduated | No |
Material | PFA |
Shape | Cylindrical |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10132901
|
KARTELL
0167200 |
30 mL |
£71.05
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10274041
|
KARTELL
0167300 |
60 mL |
£85.35
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10204191
|
KARTELL
0167500 |
180 mL |
£111.00
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- With screw cap (PFA) for sample collection, transport and storage.
- PFA trace analysis labware maintains its stability at temperatures from -200°C to
+260°C (-328°F to +500°F). - PFA is manufactured without metals such as calcium, aluminum, iron, magnesium, nickel, copper, manganese or zinc. These metals can leach into samples from other containers, and contaminate samples.
- PFA is inert with most reagents, including nitric and hydrochloric acid routinely used in cleaning trace analysis labware. This stability significantly reduces the risk of cross-contamination.
- PFA containers often extend the stability of low (ppb) concentration standards. Such stability reduces the time and cost of renewing trace analysis standard solutions.
- Translucency is important for making accurate measurements using volumetric labware.
- Conventional labware for trace analysis requires time-consuming and expensive cleaning. The hydrophobic and antiadhesive properties of the exceptionally smooth-surfaced PFA labware simplify the cleaning process, making it ideal for trace analysis.
Specifications
Specimen Container | |
No | |
Cylindrical | |
Screw Cap |
Yes | |
PFA | |
Non-sterile | |
Certified Quality |