missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Stainless steel beaker

Beaker with scraper, contents 1000 ml. Ideal for powder, granules, pastes, mud.

Brand:  Buerkle™ 5354-2000

201.21 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10575922

  • £221.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sterilisable
  • Stainless steel AISI 304 (1.4301)
  • For industry and water treatment
Specifications

Specifications

Stainless Steel
1000 mL
Pendulum
For Industry and Water Treatment
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ Stainless steel beaker > Material: Stainless Steel; Capacity: 1000mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.