missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh, Fisher Chemical™
£74.80 - £170.00
Chemical Identifiers
CAS | 14808-60-7 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10336100
|
Fisher Chemical
S/0360/60 |
1 kg |
£74.80
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10519770
|
Fisher Chemical
S/0360/63 |
3 kg |
£170.00
3kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh
Chemical Identifiers
14808-60-7 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
dioxosilane |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 | |
O=[Si]=O |
Specifications
2230°C | |
14808-60-7 | |
Solid | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08g/mol |
1610°C | |
Beige | |
O2Si | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
Plastic powder jar |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh, Fisher Chemical™