missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sand, White Quartz, Honeywell Fluka™
Purum p.a., powder
£73.74 - £287.29
Chemical Identifiers
CAS | 14808-60-7 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15697260
|
Honeywell-Fluka
83340-1KG |
1 kg | Poly bottle |
£73.74
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15607270
|
Honeywell-Fluka
83340-5KG |
5 kg | Poly bottle |
£287.29
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
14808-60-7 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
dioxosilane |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 | |
O=[Si]=O |
Specifications
2210°C | |
14808-60-7 | |
Solid | |
SiO2 | |
NONH for all modes of transport | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08g/mol | |
Sand, white quartz |
1710°C | |
White | |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
Odorless |
Safety and Handling
P314
missing translation for 'rtecsNumber' : 238-878-4
Product Identifier
- Sand, white quartz