missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Sanitary PVDF Clamps
Marque: Masterflex™ 31811-03
Code : NEW
Informations supplémentaires : Weight : 0.50000kg
Spécification
Autoclavable | |
Clamp | |
Sanitary Fitting | |
138°C |
1 Ea. | |
PVDF | |
63.5 mm |