missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap
For use in RT/Quantitative-PCR. Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap are comprehensive thermal cycler compatible.
Brand: Greiner Bio-One 682201
This item is not returnable.
View return policy
Description
- Thin-walled for heat transfer during amplification
- Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
- Packaging labelled with product symbols for warehousing
- Used for sample handling

Specifications
Natural | |
Non-sterile | |
Flat Cap |
0.5 mL | |
Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap > Vol.: 0.5mL; Natural
Spot an opportunity for improvement?Share a Content Correction