Learn More
Sartorius Sartoclear Dynamics™ Lab V Filtration Kit
Sartorius Sartoclear Dynamics™ Lab V Kits designed for the clarification and sterile filtration of mammalian cell culture with diatomaceous earth as filter aid.
£246.00 - £1055.00
Product Code | Brand | Capacity | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Includes | Price | Quantity & Availability | ||||
17494422
|
Sartorius
SDLV-0150-02E0-E |
150 mL | 1 x 180E02---------E (12 x Sartolab™ RF 150; 0.22 μm; PES; single-packaged; sterile); 1 x SDLKG-01.0------2 (24 x pouches of filter aid, 2 g) |
£246.00
Pack of 12 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
17404432
|
Sartorius
SDLV-0150-05E0-2 |
150 mL | 2 x 180E02---------E (24 x Sartolab™ RF 150; 0.22 μm; PES; single-packaged; sterile); 1 x SDLKG-05.0------2 (24 x pouches of filter aid, 5 g) |
£530.00
Pack of 24 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
17414432
|
Sartorius
SDLV-0250-05E0-2 |
250 mL | 2 x 180E03---------E (24 x Sartolab™ RF 250; 0.22 μm; PES, single-packaged; sterile); 1 x SDLKG-05.0------2 (24 x pouches of filter aid, 5 g) |
£570.00
Pack of 24 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
17424432
|
Sartorius
SDLV-0250-10E0-2 |
250 mL | 2 x 180E03---------E (24 x Sartolab™ RF 250; 0.22 μm; PES; single-packaged; sterile); 1 x SDLKG-10.0------2 (24 x pouches of filter aid, 10 g) |
£601.00
Pack of 24 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
17434432
|
Sartorius
SDLV-0500-05E0-2 |
500 mL | 2 x 180E04---------E (24 x Sartolab™ RF 500; 0.22 μm; PES; single-packaged; sterile); 1 x SDLKG-05.0------2 (24 x pouches of filter aid, 5 g) |
£720.00
Pack of 24 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
17444432
|
Sartorius
SDLV-0500-10E0-2 |
500 mL | 2 x 180E04---------E (24 x Sartolab™ RF 500; 0.22 μm; PES, single-packaged; sterile); 1 x SDLKG-10.0------2 (24 x pouches of filter aid, 10 g) |
£795.00
Pack of 24 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
17454432
|
Sartorius
SDLV-0500-20E0-E |
500 mL | 1 x 180E04---------E (12 x Sartolab™ RF 500; 0.22 μm: PES; single-packaged; sterile); 1 x SDLKG-20.0-----E (Containing 1 box of 12 x 20 g DE filter aid pouches) |
£419.00
Pack of 12 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
17464432
|
Sartorius
SDLV-1000-10E0-2 |
1000 mL | 2 x 180E05---------E (24 x Sartolab™ RF 1000; 0.22 μm: PES; single-packaged; sterile); 1 x SDLKG-10.0------2 (24 x pouches of filter aid, 10 g) |
£1055.00
Pack of 24 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
17474432
|
Sartorius
SDLV-1000-20E0-E |
1000 mL | 1 x 180E05---------E (12 x Sartolab™ RF 1000; 0.22μm; PES; single-packaged; sterile); 1 x SDLKG-20.0-----E (Containing 1 box of 12 x 20 g DE filter aid pouches) |
£555.00
Pack of 12 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
17484432
|
Sartorius
SDLV-1000-40E0-E |
1000 mL | 1 x 180E05---------E (12 x Sartolab™ RF 1000; 0.22 μm; PES; single-packaged; sterile); 1 x SDLKG-20.0-----E (Containing 2 boxes of 12 x 20 g DE filter aid pouches) |
£621.00
Pack of 12 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
Description
Comprised of:
- A graduated funnel with a polyethersulfone (PES) membrane, a vacuum adapter and a lid
- A bottle, with graduation and writing field
- A tube connector for vacuum connection (for stand-alone filtration)
- A screw cap for storage of the filtrate (individually wrapped to maintain sterility) history
Benefits:
- Kits for a single-step mammalian cell culture harvesting
- Ready to use
- Fast and effortless
Specifications
Vacuum Filtration Units |