missing translation for 'onlineSavingsMsg'
Learn More

Sartorius arium™ advance EDI Electodeionization Water System, ASTM Type 2

Outfit your lab with he Sartorisu™ arium™ advance EDI provide consistently high quality Type 2 water.

Brand:  Sartorius H2O-EDI-2-T


4975.55 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.



Code : NEW

Additional Details : Weight : 26.00000kg

 View more versions of this product

Product Code. 14831382

  • £7750.00 / Each
In Stock
to see stock.
Add to basket

Description

Description

The arium™ advance EDI pure water system delivers Type II water with the best quality at rates of up to 10l/hr. The system attains the highest retention rates of ions along with optimal water yield, and reliably removes oxidants, heavy metal ions and particulates from feed water. The pure water produced is stored in arium™ bagtank systems. If a Sartopore™ 2 150 final filter is used in the system, the water is practically free of microorganisms.

  • Innovative iJust technology
  • Uses unique bagtank technology with dispense gun connections
  • User-friendly, easy operation
  • Unique pre-filter technology with activated carbon, phosphate-free catalyst and 5μm particle filter

Benefits:

  • Optimized water consumption
  • Meets customer's lab requirements and saves maintenance time
  • No user manual needed, intuitive operation of the menu
  • Highly efficient reduces colloids and chlorine, oxidizing agents, organic and water-soluble heavy metals

Applications:

  • Microbiological media and reagents
  • Solutions for chemical analysis and synthesis
  • Histology
  • ELISA, RIA
  • Buffers and pH solutions
  • Feed water for laboratory devices, such as autoclaves, glassware washers, humidifiers, water baths etc.

Specifications

Specifications

0.2 to 0.07 Product water, Less than1500 feed water
Greater than99%
<10
Type 2
<50 product water, <1000 feed water
35.6°F to 86°F
Feed water for laboratory devices, such as autoclaves, Glassware washers, humidifiers, water baths etc
Benchtop
4 to 10
35.6°F to 95°F
4
13.7 in.
19.7 in.
17.75 in.
57.3 lb.
100/240 V 50/60 Hz
50/60 Hz
Greater than99%
<1 CFU/L
10 L/hr.
<5 to 15 product water
EDI Electrodeionization Water System
2°C to 30°C
80%
<1/mL
5°C to 45°C
2°C to 35°C
<1
35 cm
50.1 cm
45.1 cm
26 kg
100/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers