missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius arium™ advance EDI Electodeionization Water System, ASTM Type 2
Outfit your lab with he Sartorisu™ arium™ advance EDI provide consistently high quality Type 2 water.
£6920.00 - £9120.00
Specifications
Conductivity | 0.2 to 0.07 Product water, Less than1500 feed water |
---|---|
Contaminant Rejection (Microbiological) | Greater than99% |
Contaminant Rejection (Organic) | Greater than99% |
Bacterial Content | <1 CFU/L |
Daily Output | <10 |
Product Code | Brand | Mounting | Flow Rate | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mounting | Flow Rate | Price | Quantity & Availability | ||||
14831382
|
Sartorius
H2O-EDI-2-T |
Benchtop | 10 L/hr. |
£7750.00
Each |
In Stock
Log in to see stock. |
||||
14821382
|
Sartorius
H2O-EDI-2-B |
Wall | 10 L/hr. |
£7750.00
Each |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||||
15307373
|
Sartorius
H2O-EDI-1-T |
Benchtop | 5 L/hr. |
£9120.00
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
15809354
|
Sartorius
H2O-EDI-1-B |
Wall | 5 L/hr. |
£6920.00
Each |
Estimated Shipment: 20-11-2024
Log in to see stock. |
||||
Description
The arium™ advance EDI pure water system delivers Type II water with the best quality at rates of up to 10l/hr. The system attains the highest retention rates of ions along with optimal water yield, and reliably removes oxidants, heavy metal ions and particulates from feed water. The pure water produced is stored in arium™ bagtank systems. If a Sartopore™ 2 150 final filter is used in the system, the water is practically free of microorganisms.
- Innovative iJust technology
- Uses unique bagtank technology with dispense gun connections
- User-friendly, easy operation
- Unique pre-filter technology with activated carbon, phosphate-free catalyst and 5μm particle filter
Benefits:
- Optimized water consumption
- Meets customer's lab requirements and saves maintenance time
- No user manual needed, intuitive operation of the menu
- Highly efficient reduces colloids and chlorine, oxidizing agents, organic and water-soluble heavy metals
Applications:
- Microbiological media and reagents
- Solutions for chemical analysis and synthesis
- Histology
- ELISA, RIA
- Buffers and pH solutions
- Feed water for laboratory devices, such as autoclaves, glassware washers, humidifiers, water baths etc.
Specifications
0.2 to 0.07 Product water, Less than1500 feed water | |
Greater than99% | |
<10 | |
<5 to 15 product water | |
35.6°F to 86°F | |
Feed water for laboratory devices, such as autoclaves, Glassware washers, humidifiers, water baths etc | |
<1/mL | |
5°C to 45°C | |
2°C to 35°C | |
<1 | |
35 cm | |
50.1 cm | |
45.1 cm | |
26 kg | |
100/240 V |
Greater than99% | |
<1 CFU/L | |
Type 2 | |
<50 product water, <1000 feed water | |
2°C to 30°C | |
80% | |
4 to 10 | |
35.6°F to 95°F | |
4 | |
13.7 in. | |
19.7 in. | |
17.75 in. | |
57.3 lb. | |
100/240 V 50/60 Hz | |
50/60 Hz |