Learn More
Sartorius Glass Microfiber Filters
Binder free glass microfiber filters are suitable for analytical and gravimetric analyses. They are also used as prefilter.
£17.10 - £167.00
Specifications
Color (Overall) | White |
---|---|
Type | Glass Microfiber Filter |
Format | Filter Disc |
For Use With (Application) | Ideal for critical analyses in which impurities can be washed out of the filter |
Material | Borosilicate Glass |
Product Code | Brand | Diameter (Metric) | Grade | Particle Retention | Thickness (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Grade | Particle Retention | Thickness (Metric) | Price | Quantity & Availability | ||
11790436
|
Sartorius
FT-3-1104-125 |
12.5 cm | MGD | 2.7 μm | 0.05 cm |
£107.00
Pack of 50 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
11717415
|
Sartorius
FT-3-1104-150 |
15 cm | MGD | 2.7 μm | 0.05 cm |
£134.00
Pack of 50 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
11770436
|
Sartorius
FT-3-1104-100 |
10 cm | MGD | 2.7 μm | 0.05 cm |
£74.80
Pack of 50 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
11707415
|
Sartorius
FT-3-1104-090 |
9 cm | MGD | 2.7 μm | 0.05 cm |
£64.00
Pack of 50 |
In Stock
Log in to see stock. |
||
11760436
|
Sartorius
FT-3-1104-070 |
7 cm | MGD | 2.7 μm | 0.05 cm |
£42.10
Pack of 50 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
11797405
|
Sartorius
FT-3-1104-047 |
4.7 cm | MGD | 2.7 μm | 0.05 cm |
£24.90
Pack of 50 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||
11750436
|
Sartorius
FT-3-1104-050 |
5 cm | MGD | 2.7 μm | 0.05 cm |
£28.70
Pack of 50 |
In Stock
Log in to see stock. |
||
10008844
|
Sartorius
FT-3-1104-013 |
1.3 cm | MGD | 2.7 μm | 0.05 cm |
£29.60
Pack of 100 |
In Stock
Log in to see stock. |
||
10357741
|
Sartorius
FT-3-1104-025 |
2.5 cm | MGD | 2.7 μm | 0.05 cm |
£17.10
Pack of 50 |
In Stock
Log in to see stock. |
||
15972860
|
Sartorius
FT-3-1102-240 |
24 cm | MGB | 1 μm | 0.07 cm |
£153.00
Pack of 25 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
11700436
|
Sartorius
FT-3-1102-150 |
15 cm | MGB | 1 μm | 0.07 cm |
£167.00
Pack of 50 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
10602602
|
Sartorius
FT-3-1102-090 |
9 cm | MGB | 1 μm | 0.07 cm |
£76.10
Pack of 50 |
In Stock
Log in to see stock. |
||
11780426
|
Sartorius
FT-3-1102-110 |
11 cm | MGB | 1 μm | 0.07 cm |
£99.90
Pack of 50 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
11750426
|
Sartorius
FT-3-1102-070 |
7 cm | MGB | 1 μm | 0.07 cm |
£59.00
Pack of 50 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
11740426
|
Sartorius
FT-3-1102-055 |
5.5 cm | MGB | 1 μm | 0.07 cm |
£43.50
Pack of 50 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
11730426
|
Sartorius
FT-3-1102-050 |
5 cm | MGB | 1 μm | 0.07 cm |
£42.10
Pack of 50 |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||
11720426
|
Sartorius
FT-3-1102-047 |
4.7 cm | MGB | 1 μm | 0.07 cm |
£39.00
Pack of 50 |
In Stock
Log in to see stock. |
||
11780416
|
Sartorius
FT-3-1101-240 |
24 cm | MGA | 1.6 μm | 0.02 cm |
£128.00
Pack of 50 |
In Stock
Log in to see stock. |
||
10573262
|
Sartorius
FT-3-1102-025 |
2.5 cm | MGB | 1 μm | 0.07 cm |
£20.50
Pack of 50 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
15962860
|
Sartorius
FT-3-1102-024 |
2.4 cm | MGB | 1 μm | 0.07 cm |
£20.20
Pack of 50 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
11790416
|
Sartorius
FT-3-1102-021 |
2.1 cm | MGB | 1 μm | 0.07 cm |
£20.20
Pack of 50 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
10203481
|
Sartorius
FT-3-1101-110 |
11 cm | MGA | 1.6 μm | 0.02 cm |
£95.40
Pack of 100 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
11795296
|
Sartorius
FT-3-1101-100 |
10 cm | MGA | 1.6 μm | 0.02 cm |
£81.10
Pack of 100 |
In Stock
Log in to see stock. |
||
10260102
|
Sartorius
FT-3-1101-080 |
8 cm | MGA | 1.6 μm | 0.02 cm |
£70.20
Pack of 100 |
In Stock
Log in to see stock. |
||
10089620
|
Sartorius
FT-3-1101-090 |
9 cm | MGA | 1.6 μm | 0.02 cm |
£75.70
Pack of 100 |
In Stock
Log in to see stock. |
||
10223381
|
Sartorius
FT-3-1101-055 |
5.5 cm | MGA | 1.6 μm | 0.02 cm |
£47.60
Pack of 100 |
In Stock
Log in to see stock. |
||
10692592
|
Sartorius
FT-3-1101-070 |
7 cm | MGA | 1.6 μm | 0.02 cm |
£64.10
Pack of 100 |
In Stock
Log in to see stock. |
||
10769412
|
Sartorius
FT-3-1101-050 |
5 cm | MGA | 1.6 μm | 0.02 cm |
£40.60
Pack of 100 |
In Stock
Log in to see stock. |
||
10707081
|
Sartorius
FT-3-1101-047 |
4.7 cm | MGA | 1.6 μm | 0.02 cm |
£45.30
Pack of 100 |
In Stock
Log in to see stock. |
||
11735296
|
Sartorius
FT-3-1101-037 |
3.7 cm | MGA | 1.6 μm | 0.02 cm |
£29.70
Pack of 100 |
In Stock
Log in to see stock. |
||
10527631
|
Sartorius
FT-3-1101-025 |
2.5 cm | MGA | 1.6 μm | 0.02 cm |
£29.60
Pack of 100 |
In Stock
Log in to see stock. |
||
Description
Binder free glass microfibre filters are made of 100 % borosilicate, are highly pure and extremely white. They continue to perform over long periods unlike cellulose filters whose rate of filtration drops off rapidly as the particulate load increases. They are ideal for critical analyses in which impurities can be washed out of the filter. Moreover, binder free glass microfibre filters are temperature resistant up to 500 or 550°C (grade MG 550-HA ).
They are available in different formats as in rolls, discs and sheets.
Specifications
White | |
Filter Disc | |
Borosilicate Glass | |
500°C or 550°C |
Glass Microfiber Filter | |
Ideal for critical analyses in which impurities can be washed out of the filter | |
High |