Learn More
Sartorius Microsart™ e.motion Membrane Filters
Membrane Filters suitable for use in the Microsart e.motion dispenser
£341.00 - £524.00
Specifications
Material Cellulose Nitrate
Sterility Sterile
View More Specs
Products 13
Product Code Brand Color Diameter (Metric) Pore Size Grid Color Price Quantity & Availability  
Product Code Brand Color Diameter (Metric) Pore Size Grid Color Price Quantity & Availability  
11760796
View Documents Promotion Details
Sartorius
13005Z-50----SCM
Gray 50 mm 0.65 μm White
£348.00
Pack of 300
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11710806
View Documents Promotion Details
Sartorius
13806Z-50----SCM
Green 50 mm 0.45 μm Dark Green
£348.00
Pack of 300
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11874232
View Documents Promotion Details
Sartorius
13004Z-47----SCM
Gray 47 mm 0.8 μm White
£348.00
Pack of 300
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11720806
View Documents Promotion Details
Sartorius
13906Z-50----SCM
White 50 mm 0.45 μm Green
£348.00
Pack of 300
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11780796
View Documents Promotion Details
Sartorius
13006Z-50----SCM
Gray 50 mm 0.45 μm White
£348.00
Pack of 300
View alternative products
Save Up to 
»
Estimated Shipment: 17-09-2024
to see stock.
Add to basket
15932460
View Documents Promotion Details
Sartorius
11402Z-47----SCM
White 47 mm 0.3 μm Black
£348.00
Pack of 300
View alternative products
Save Up to 
»
In Stock
to see stock.
11720796
View Documents Promotion Details
Sartorius
11406Z-50----SCM
White 50 mm 0.45 μm Black
£348.00
Pack of 300
View alternative products
Save Up to 
»
Estimated Shipment: 17-09-2024
to see stock.
Add to basket
10204662
View Documents Promotion Details
Sartorius
11406Z-47----SCM
White 47 mm 0.45 μm Black
£524.00
Pack of 300
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11814232
View Documents Promotion Details
Sartorius
11407Z-47----SCM
White 47 mm 0.2 μm Black
£444.00
Pack of 300
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10478072
View Documents Promotion Details
Sartorius
13006Z-47----SCM
Gray 47 mm 0.45 μm White
£348.00
Pack of 300
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10682353
View Documents Promotion Details
Sartorius
13806Z-47----SCM
Dark Green 47 mm 0.45 μm Green
£348.00
Pack of 300
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10132132
View Documents Promotion Details
Sartorius
13906Z-47----SCM
White 47 mm 0.45 μm Green
£348.00
Pack of 300
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11750796
View Documents Promotion Details
Sartorius
13004Z-50----SCM
Gray 50 mm 0.8 μm White This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Membrane Filters suitable for use in the Microsart e.motion dispenser
The membrane filters suitable for use in the Microsart™ e.motion dispenser are sterile-sealed, without protective paper on top of each filter, in a specially designed individual package on a band. The special pleating of the band of membrane filter units ensures that they are perfectly flat when dispensed.The shape of the sealed band guarantees uniform dispensing of the individual membrane filters.The Microsart™ e.motion Membrane Filters are also ideal suitable for all users of Sartorius Nutrient Pad Sets: Most of the NPS types are available with membranes for dispensers.
  • outstanding recovery rates for microorganisms
  • 0.45μm are acc. to ISO 7704
  • Multi-fit: Fits into other dispensers
  • protective paper-free
  • packaged on a special pleated band
  • product data are printed on
  • High Flow membranes available
  • gamma irradiated, 25kGray
Design
47 or 50mm in diameter, white, grey or green and gridded
Growth Promotion Test acc. to ISO 7704
  • no enhancement or inhibition by the grid lines
  • no enhancement or inhibition due to chemical extractables
  • no enhancement or inhibition by the sterilization process
Specifications
Specifications
Cellulose Nitrate
Sterile