Learn More
Sartorius Traditional Vacuum Pumps
Neoprene membrane pumps with low noise level, oil- and maintenance-free and is a reliable vacuum
Brand:  Sartorius 16695
 View more versions of this product
Product Code. 15972540
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The Microsart™ e.jet vacuum laboratory pump is designed to create sufficient vacuum for filtration and unlike traditional vacuum pumps, it allows simultaneous transfer of the filtered liquid to waste. The Microsart™ e.jet Transfer Pump is ideal for sample filtration in Microbiology achieving a trans membrane pressure below 700 mbar in compliance with ISO 8199 (Water quality - General guidance on the enumeration of micro-organisms by culture). Constant flow rates and a defined maximum vacuum guarantee smooth and reliable filtration. The Microsart™ e.jet Transfer Pump greatly reduces operating complexity and is an ideal accessory for 3-branch and 1-branch manifolds.
The Microsart™ e.jet is designed to pump both gas and liquids, meaning no loss of efficiency or malfunctions from water drawn into the pump head. The vacuum pump series Microsart™ maxi.vac and Microsart™ mini.vac provide up to date technology for daily use in the Microbiology laboratory environment. They are electric neoprene membrane pumps and therefore oil- and maintenance-free with a low noise level. The vacuum produced by these pumps is controlled and can be easily adjusted to your specifications. Thus sensitive cells (e.g. bacteria) are concentrated on the surface of a membrane filter under better conditions, which results in decreased mortality of the cells, higher recovery rates and shorter incubation times.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 mbar
110 V 60 Hz
18.7 cm
9.9 lb.
104°F
20 L/min.
60 Hz
14.5 cm
203 mm
145 mm
110 V
0.75 A
7.3 x 8 x 5.7 in. (18.7 x 20.3 x 14.5 cm)
80 w
4.5 kg
40°C
Combisart
7.4 in.
8 in.
8 in.
Single filtration run with individual filter station for microbial enumeration