missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivacon™ 2 DNA Concentrator

Centrifugal units for fast DNA concentration and buffer exchange of 2 ml samples

Brand:  Sartorius VN02H31

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11744905

  • £205.00 / Pack of 25
Estimated Shipment: 20-09-2024
to see stock.
Add to basket

Description

Description

Vivacon 2 are centrifugal devices for DNA concentration and buffer exchange applications for samples up to 2 ml. For optimal performance with very diluted samples, e.g. forensic specimens, Vivacon 2 is equipped with patented Hydrosart™ regenerated cellulose membranes with extremely low unspecific binding properties and high flux. High recoveries and excellent reproducibility are paired with convenience offered by volume graduation and molecular weight cut-offs printed on individual devices. Vivacon 2 features a horizontal membrane design optimal for DNA fractionation (primer removal) as well as protein fractionation (peptide removal). The possibility of a reverse spin after sample processing assures complete concentrate recovery which is especially important when working with low sample concentrations.

  • Convenient handling in fixed-angle rotors
  • Low binding material
  • Reverse spin possibility
  • Graduation and MWCO printed on device
  • Complete and highly reproducible concentrate recovery

Specifications

Specifications

50ng/mL
Polycarbonate Body, Polypropylene Filtrate Vessel, Polypropylene Back Spin Vial, Polypropylene Concentrator Cap, Hydrosart Membrane, Silicone O-ring
0.95 cm2
2500xG
DNA, peptide
2 mL
4.92 x 0.62 in. (12.5 x 1.6cm)
4.52 in.
11.5 cm
1.6 cm
45 min.
50,000
DNA concentration, desalting, buffer exchange, fractionation
25 Pack
Vivacon 2 DNA Concentrator
10 μL
4.92 in.
12.5 cm
0.62 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers