missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivacon™ 2 DNA Concentrator
BIOPHARMA

Centrifugal units for fast DNA concentration and buffer exchange of 2 ml samples

£207.00 - £3100.00

Specifications

Concentration 50ng/mL
Material Polycarbonate Body, Polypropylene Filtrate Vessel, Polypropylene Back Spin Vial, Polypropylene Concentrator Cap, Hydrosart Membrane, Silicone O-ring
Filtration Area 0.95 cm2
For Use With (Application) DNA concentration, desalting, buffer exchange, fractionation
Sample Type DNA, peptide
View More Specs

Products 11
Product Code Brand MWCO Quantity Price Quantity & Availability  
Product Code Brand MWCO Quantity Price Quantity & Availability  
11726355
BIOPHARMA
View Documents
Sartorius
VN02H02
10 kDa 100 Pack
£761.00
Pack of 100
In Stock
to see stock.
Add to basket
 
12313770
BIOPHARMA
View Documents
Sartorius
VN02H21
30 kDa 25 Pack
£207.00
Pack of 25
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
11736355
BIOPHARMA
View Documents
Sartorius
VN02H22
30 kDa 100 Pack
£753.00
Pack of 100
In Stock
to see stock.
Add to basket
 
11760846
View Documents
Sartorius
VN02H23
30 kDa 500 Pack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11744905
BIOPHARMA
View Documents
Sartorius
VN02H31
50,000 25 Pack
£207.00
Pack of 25
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
11759255
BIOPHARMA
View Documents
Sartorius
VN02H32
50,000 100 Pack
£743.00
Pack of 100
In Stock
to see stock.
Add to basket
 
11779255
BIOPHARMA
View Documents
Sartorius
VN02H42
100,000 100 Pack
£753.00
Pack of 100
In Stock
to see stock.
Add to basket
 
11704905
BIOPHARMA
View Documents
Sartorius
VN02H91
2000 25 Pack
£219.00
Pack of 25
In Stock
to see stock.
Add to basket
 
11780846
BIOPHARMA
View Documents
Sartorius
VN02H43
100,000 500 Pack
£3100.00
Pack of 500
In Stock
to see stock.
Add to basket
 
11769255
BIOPHARMA
View Documents
Sartorius
VN02H41
100,000 25 Pack
£220.00
Pack of 25
In Stock
to see stock.
Add to basket
 
11760706
BIOPHARMA
View Documents
Sartorius
VN02H92
2000 100 Pack
£753.00
Pack of 100
In Stock
to see stock.
Add to basket
 
Description

Description

Vivacon 2 are centrifugal devices for DNA concentration and buffer exchange applications for samples up to 2 ml. For optimal performance with very diluted samples, e.g. forensic specimens, Vivacon 2 is equipped with patented Hydrosart™ regenerated cellulose membranes with extremely low unspecific binding properties and high flux. High recoveries and excellent reproducibility are paired with convenience offered by volume graduation and molecular weight cut-offs printed on individual devices. Vivacon 2 features a horizontal membrane design optimal for DNA fractionation (primer removal) as well as protein fractionation (peptide removal). The possibility of a reverse spin after sample processing assures complete concentrate recovery which is especially important when working with low sample concentrations.

  • Convenient handling in fixed-angle rotors
  • Low binding material
  • Reverse spin possibility
  • Graduation and MWCO printed on device
  • Complete and highly reproducible concentrate recovery

Specifications

Specifications

50ng/mL
0.95 cm2
DNA, peptide
2 mL
4.92 x 0.62 in. (12.5 x 1.6cm)
4.52 in.
11.5 cm
1.6 cm
Polycarbonate Body, Polypropylene Filtrate Vessel, Polypropylene Back Spin Vial, Polypropylene Concentrator Cap, Hydrosart Membrane, Silicone O-ring
DNA concentration, desalting, buffer exchange, fractionation
Vivacon 2 DNA Concentrator
10 μL
4.92 in.
12.5 cm
0.62 in.
Videos
SDS
Documents

Documents

Product Certifications