Learn More
Sartorius Vivapure™ AdenoPACK 100 Kit
Fast purification of up to 1 x 1013 viral particles
Brand: Sartorius VS-AVPQ101
817.00 GBP valid until 2024-12-31
Use promo code "21963" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.25000kg
Description
Viral particles Vivapure AdenoPACK 100 is optimally suited for adenovirus purification from up to 200 ml cell culture for in vitro transfection. This flexible kit contains two AdenoPACK 100 units, which can be either used in tandem for the purification of up to 200 ml cell culture supernatant or individually for purifying virus from up to 60 ml cell culture. The purification is performed manually with a syringe optionally attached to a lab stand. However, for even more convenience, protocols are provided for optionally running the virus purification with a peristaltic pump or with an infusion pump, in additional to detailed instructions for a manual operation supplied with the Kit. The accessories needed for the operation with a pump are supplied as individual products.
High-speed process: completed in 2 hours
High virus recoveries
Convenient, easy to use
For processing 20-200 ml cell supernatants
- High-speed process: completed in 2 hours High virus recoveries Convenient, easy to use For processing 20-200 ml cell supernatants
Specifications
<0.025 EU/mL | |
20 to 200 mL of Adenovirus supernatants | |
-20°C |