Learn More
Sartorius Vivapure™ AdenoPACK 500 Kit
Pump driven Kit for larger volumes
Brand: Sartorius VS-AVPQ501
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Vivapure™ AdenoPACK 500 is the direct upscale kit to the AdenoPACK 100, for adenovirus purification. In only 2 hours up to 3 × 1013 adenovirus particles are purified and concentrated from 500 ml cell culture. This completely ready-to-use kit is conveniently operated by a laboratory pump, offering optimal flow control and minimal hands-on time. This easy to use product replaces lengthy and inefficient cesium chloride density gradient methods.
- Pump driven kit
- Easy and controlled purification of Adenovirus from large cell culture volumes
- Virus purification based on membrane adsorber technology
- High-speed process completed in about 2 hours
- Convenient, easy to use alternative to CsCl density gradients
- Able to process 10 to 25 plates (up to 500 ml)
- Flexible, most cost-effective kit in the market for purifying 1-3 × 1013 viral particles
Specifications
<0.025 EU/mL | |
500 mL of Adenovirus supernatants | |
-20°C |