Learn More
Sartorius Vivapure™ LentiSELECT 1000 Purification and Concentration Kit
Fast purification of up to 3 x109 viral particles
Brand: Sartorius VS-LVPQ1000
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Vivapure™ LentiSELECT 1000 is a pump driven kit for VSV-G pseudotyped lentivirus purification. The rapid 6 hour protocol results in a recovery of 4–5 x 109 infective particles per ml (total volume 2mL) from 1000mL cell culture supernatant.
This kit is to be operated by a laboratory pump and contains all necessary buffers and ultrafiltration devices for optimal convenience. The traditional time consuming ultracentrifugation method is replaced by this fast and simple Vivapure LentiSELECT 1000 kit.
Typically, for a normal yielding vector, a 1000mL cell culture purified using this method will yield up to 4–5 x 109 infective particles in 1 ml (total volume 2mL).
- Fast
- Non-toxic
- Better purity than pelleting by ultracentrifugation
- Convienient
- Cost effective
Specifications
1000 mL of Lentivirus supernatants |