Learn More
Sartorius Vivapure™ LentiSELECT 500 Purification and Concentration Kit
BIOPHARMA
Fast purification of up to 3 x109 viral particles
Brand:  Sartorius VS-LVPQ500
1170.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11740865
  • £1600.00 / Each
Estimated Shipment: 09-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Vivapure™ LentiSELECT 500 is optimally suited for VSV-G pseudotyped lentivirus purification from up to 500mL cell culture and contains all reagents and devices necessary for purifying up to 2-5 x 109 infective particles.
The whole purification procedure is simply operated by a laboratory pump, which min imizes hands-on time. Unlike conventional purification methods as ultracentrifugation, Vivapure LentiSELECT 500 offers a fast and simple solution for purifying VSV-G pseudotyped lentiviruses making expensive purification equipment like ultracentrifuges redundant.
Typically, for a normal yielding vector, a 500mL cell culture purified using this method will yield up to 2-5 x 109 infective particles in 1mL (total volume 1mL).
  • Fast
  • Non-toxic
  • Better purity than pelleting by ultracentrifugation
  • Convienient
  • Cost effective
TRUSTED_SUSTAINABILITY
Specifications
Specifications
500 mL of Lentivirus
40 to 1000 mL Cell Cultures
VSV-G Pseudotyped Lentiviral Vectors
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius Vivapure™ LentiSELECT 500 Purification and Concentration Kit > LentiSELECT 500; Vivaspin 20 300kDa (2)